Assay Details
Gene Symbol: Sgcz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004867638.2 | XP_004867695.1 | 6 - 7 | 1398 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
