Assay Details
| Gene Symbol: Lrrc39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004869124.2 | XP_004869181.1 | 7 - 8 | 1036 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrrc39| Gene Symbol: Lrrc39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004869124.2 | XP_004869181.1 | 7 - 8 | 1036 | 63 | |
Set Membership: |
|