Assay Details
| Gene Symbol: Stam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004859965.2 | XP_004860022.1 | 2 - 3 | 314 | 65 | |
| XM_013077868.2 | XP_012933322.1 | 2 - 3 | 173 | 65 | ||
| XM_021240383.1 | XP_021096042.1 | 3 - 4 | 361 | 65 | ||
| XM_021240384.1 | XP_021096043.1 | 2 - 3 | 101 | 65 | ||
More Information
Set Membership: |
|

Stam
Genomic Map