Assay Details
| Gene Symbol: Papss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021245399.1 | XP_021101058.1 | 7 - 8 | 855 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Papss1| Gene Symbol: Papss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021245399.1 | XP_021101058.1 | 7 - 8 | 855 | 66 | |
Set Membership: |
|