Assay Details
| Gene Symbol: Mrpl55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004874720.3 | XP_004874777.1 | 3 - 4 | 403 | 105 | |
| XM_004874721.2 | XP_004874778.1 | 3 - 4 | 381 | 105 | ||
| XM_004874722.3 | XP_004874779.1 | 3 - 4 | 311 | 105 | ||
| XM_004874723.2 | XP_004874780.1 | 3 - 4 | 346 | 105 | ||
| XM_013069252.2 | XP_012924706.1 | 3 - 4 | 290 | 105 | ||
| XM_021252104.1 | XP_021107763.1 | 3 - 4 | 638 | 105 | ||
| XM_021252105.1 | XP_021107764.1 | 3 - 4 | 358 | 105 | ||
More Information
Set Membership: |
|

Mrpl55
Genomic Map