Assay Details
| Gene Symbol: Pter | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004859978.3 | XP_004860035.1 | 2 - 3 | 973 | 78 | |
| XM_004859979.3 | XP_004860036.1 | 2 - 3 | 581 | 78 | ||
| XM_004859981.3 | XP_004860038.1 | 2 - 3 | 509 | 78 | ||
| XM_021240446.1 | XP_021096105.1 | 2 - 3 | 624 | 78 | ||
| XM_021240447.1 | XP_021096106.1 | 2 - 3 | 568 | 78 | ||
More Information
Set Membership: |
|

Pter
Genomic Map