Assay Details
| Gene Symbol: Crybg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855440.3 | XP_004855497.1 | 15 - 16 | 5255 | 64 | |
| XM_021236886.1 | XP_021092545.1 | 15 - 16 | 5662 | 64 | ||
| XM_021236887.1 | XP_021092546.1 | 14 - 15 | 5253 | 64 | ||
| XM_021236888.1 | XP_021092547.1 | 14 - 15 | 5329 | 64 | ||
More Information
Set Membership: |
|

Crybg1
Genomic Map