Assay Details
| Gene Symbol: Lrr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004835596.3 | XP_004835653.1 | 3 - 4 | 1617 | 115 | |
| XM_021262656.1 | XP_021118315.1 | 3 - 4 | 1620 | 115 | ||
| XM_021262666.1 | XP_021118325.1 | 2 - 3 | 1518 | 115 | ||
More Information
Set Membership: |
|

Lrr1
Genomic Map