Assay Details
| Gene Symbol: Gna11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865824.2 | XP_004865881.1 | 3 - 4 | 494 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gna11| Gene Symbol: Gna11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865824.2 | XP_004865881.1 | 3 - 4 | 494 | 62 | |
Set Membership: |
|