Assay Details
| Gene Symbol: Ppt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013072959.2 | XP_012928413.1 | 7 - 8 | 1084 | 68 | |
| XM_013072960.2 | XP_012928414.1 | 7 - 8 | 864 | 68 | ||
| XM_013072961.2 | XP_012928415.1 | 7 - 8 | 816 | 68 | ||
More Information
Set Membership: |
|

Ppt2
Genomic Map