Assay Details
| Gene Symbol: Lrrc46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021239978.1 | XP_021095637.1 | 7 - 8 | 669 | 69 | |
| XM_021239979.1 | XP_021095638.1 | 7 - 8 | 636 | 69 | ||
| XM_021239980.1 | XP_021095639.1 | 6 - 7 | 730 | 69 | ||
| XM_021239981.1 | XP_021095640.1 | 6 - 7 | 691 | 69 | ||
| XM_021239982.1 | XP_021095641.1 | 6 - 7 | 684 | 69 | ||
More Information
Set Membership: |
|

Lrrc46
Genomic Map