Assay Details
| Gene Symbol: Uck1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004849090.3 | XP_004849147.1 | 2 - 3 | 341 | 66 | |
| XM_021261862.1 | XP_021117521.1 | 2 - 3 | 314 | 66 | ||
| XM_021261863.1 | XP_021117522.1 | 2 - 3 | 341 | 66 | ||
| XM_021261864.1 | XP_021117523.1 | 2 - 3 | 341 | 66 | ||
| XM_021261865.1 | XP_021117524.1 | 2 - 3 | 341 | 66 | ||
More Information
Set Membership: |
|

Uck1
Genomic Map