Assay Details
| Gene Symbol: LOC101697635 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004853902.3 | XP_004853959.1 | 11 - 12 | 1162 | 88 | |
| XM_013075500.2 | XP_012930954.1 | 11 - 12 | 1168 | 88 | ||
| XM_021265810.1 | XP_021121469.1 | 11 - 12 | 1170 | 88 | ||
More Information
Set Membership: |
|

LOC101697635
Genomic Map