Assay Details
| Gene Symbol: LOC101697635 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004853902.3 | XP_004853959.1 | 1 - 2 | 197 | 99 | |
| XM_013075500.2 | XP_012930954.1 | 1 - 2 | 197 | 99 | ||
| XM_013075501.2 | XP_012930955.1 | 1 - 2 | 197 | 99 | ||
| XM_021265810.1 | XP_021121469.1 | 1 - 2 | 199 | 99 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004853902.3, XM_013075500.2, XM_013075501.2, XM_021265810.1 transcripts.
Set Membership: |
|

LOC101697635
Genomic Map