Assay Details
| Gene Symbol: Arse | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867326.2 | XP_004867383.1 | 10 - 11 | 1565 | 102 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arse| Gene Symbol: Arse | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867326.2 | XP_004867383.1 | 10 - 11 | 1565 | 102 | |
Set Membership: |
|