Assay Details
| Gene Symbol: LOC101717279 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004864780.2 | XP_004864837.2 | 27 - 28 | 2831 | 85 | |
| XM_021244362.1 | XP_021100021.1 | 29 - 30 | 3086 | 85 | ||
| XM_021244363.1 | XP_021100022.1 | 29 - 30 | 2987 | 85 | ||
More Information
Set Membership: |
|

LOC101717279
Genomic Map