Assay Details
| Gene Symbol: Nfatc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004861916.2 | XP_004861973.1 | 4 - 5 | 1848 | 59 | |
| XM_004861917.2 | XP_004861974.1 | 4 - 5 | 1638 | 59 | ||
| XM_004861918.3 | XP_004861975.1 | 4 - 5 | 1845 | 59 | ||
| XM_021242140.1 | XP_021097799.1 | 4 - 5 | 1551 | 59 | ||
| XM_021242141.1 | XP_021097800.1 | 4 - 5 | 1637 | 59 | ||
More Information
Set Membership: |
|

Nfatc1
Genomic Map