Assay Details
Gene Symbol: Kdelc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004858569.3 | XP_004858626.1 | 5 - 6 | 1000 | 64 | |
XM_021239339.1 | XP_021094998.1 | 5 - 6 | 841 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
