Assay Details
| Gene Symbol: Zfand2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840340.2 | XP_004840397.1 | 3 - 4 | 420 | 65 | |
| XM_021254625.1 | XP_021110284.1 | 3 - 4 | 423 | 65 | ||
More Information
Set Membership: |
|

Zfand2a
Genomic Map