Assay Details
| Gene Symbol: Men1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004852455.2 | XP_004852512.1 | 9 - 10 | 1417 | 53 | |
| XM_004852456.2 | XP_004852513.1 | 9 - 10 | 1421 | 53 | ||
| XM_004852457.2 | XP_004852514.1 | 10 - 11 | 1328 | 53 | ||
| XM_021264634.1 | XP_021120293.1 | 8 - 9 | 1288 | 53 | ||
More Information
Set Membership: |
|

Men1
Genomic Map