Assay Details
Gene Symbol: Bicd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004870074.3 | XP_004870131.1 | 6 - 7 | 2429 | 59 | |
XM_004870075.3 | XP_004870132.1 | 6 - 7 | 2427 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
