Assay Details
| Gene Symbol: Kiaa0319l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004850922.3 | XP_004850979.2 | 7 - 8 | 1344 | 61 | |
| XM_004850923.3 | XP_004850980.2 | 7 - 8 | 1252 | 61 | ||
| XM_004850924.2 | XP_004850981.1 | 7 - 8 | 1440 | 61 | ||
| XM_013074339.2 | XP_012929793.1 | 7 - 8 | 1440 | 61 | ||
More Information
Set Membership: |
|

Kiaa0319l
Genomic Map