Assay Details
Gene Symbol: Hdac11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004859990.2 | XP_004860047.1 | 7 - 8 | 641 | 70 | |
XM_021240450.1 | XP_021096109.1 | 8 - 9 | 1137 | 70 | ||
XM_021240451.1 | XP_021096110.1 | 8 - 9 | 881 | 70 | ||
XM_021240452.1 | XP_021096111.1 | 6 - 7 | 1029 | 70 | ||
XM_021240453.1 | XP_021096112.1 | 6 - 7 | 610 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
