Assay Details
| Gene Symbol: Prkag1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004863051.3 | XP_004863108.1 | 5 - 6 | 317 | 79 | |
| XM_004863052.3 | XP_004863109.1 | 5 - 6 | 396 | 79 | ||
More Information
Set Membership: |
|

Prkag1
Genomic Map