Assay Details
| Gene Symbol: Sec11c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004858417.3 | XP_004858474.1 | 2 - 3 | 317 | 56 | |
| XM_004858418.3 | XP_004858475.1 | 3 - 4 | 429 | 56 | ||
| XM_004858419.3 | XP_004858476.1 | 3 - 4 | 433 | 56 | ||
More Information
Set Membership: |
|

Sec11c
Genomic Map