Assay Details
| Gene Symbol: Klf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004851151.2 | XP_004851208.1 | 1 - 2 | 545 | 77 | |
| XM_004851152.2 | XP_004851209.1 | 1 - 2 | 545 | 77 | ||
More Information
Set Membership: |
|

Klf7
Genomic Map