Assay Details
| Gene Symbol: Calcoco2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013077734.2 | XP_012933188.1 | 10 - 11 | 1279 | 60 | |
| XM_013077735.2 | XP_012933189.1 | 10 - 11 | 1273 | 60 | ||
| XM_013077736.2 | XP_012933190.1 | 10 - 11 | 1088 | 60 | ||
| XM_021239928.1 | XP_021095587.1 | 10 - 11 | 1033 | 60 | ||
More Information
Set Membership: |
|

Calcoco2
Genomic Map