Assay Details
Gene Symbol: Fam57a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004857105.3 | XP_004857162.1 | 1 - 2 | 218 | 57 | |
XM_021238149.1 | XP_021093808.1 | 1 - 2 | 218 | 57 | ||
XM_021238150.1 | XP_021093809.1 | 1 - 2 | 218 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
