Assay Details
| Gene Symbol: Leprot | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004841513.2 | XP_004841570.1 | 2 - 3 | 154 | 73 | |
| XM_021255587.1 | XP_021111246.1 | 2 - 3 | 154 | 73 | ||
More Information
Set Membership: |
|

Leprot
Genomic Map