Assay Details
| Gene Symbol: Acot11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004874628.3 | XP_004874685.1 | 6 - 7 | 955 | 53 | |
| XM_004874629.3 | XP_004874686.1 | 5 - 6 | 667 | 53 | ||
| XM_004874630.3 | XP_004874687.1 | 5 - 6 | 809 | 53 | ||
| XM_013069203.2 | XP_012924657.1 | 5 - 6 | 544 | 53 | ||
| XM_013069205.2 | XP_012924659.1 | 5 - 6 | 456 | 53 | ||
More Information
Set Membership: |
|

Acot11
Genomic Map