Assay Details
| Gene Symbol: Fmo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004853454.2 | XP_004853511.1 | 4 - 5 | 601 | 65 | |
| XM_021265456.1 | XP_021121115.1 | 3 - 4 | 483 | 65 | ||
More Information
Set Membership: |
|

Fmo2
Genomic Map