Assay Details
| Gene Symbol: Actr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004844393.3 | XP_004844450.1 | 10 - 11 | 1154 | 59 | |
| XM_013072121.2 | XP_012927575.1 | 10 - 11 | 995 | 59 | ||
| XM_013072122.2 | XP_012927576.1 | 10 - 11 | 1661 | 59 | ||
| XM_021258040.1 | XP_021113699.1 | 9 - 10 | 1030 | 59 | ||
| XM_021258041.1 | XP_021113700.1 | 10 - 11 | 996 | 59 | ||
More Information
Set Membership: |
|

Actr1b
Genomic Map