Assay Details
| Gene Symbol: Sh2d2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021249496.1 | XP_021105155.1 | 2 - 3 | 663 | 65 | |
| XM_021249497.1 | XP_021105156.1 | 2 - 3 | 662 | 65 | ||
| XM_021249498.1 | XP_021105157.1 | 2 - 3 | 662 | 65 | ||
| XM_021249499.1 | XP_021105158.1 | 2 - 3 | 661 | 65 | ||
| XM_021249500.1 | XP_021105159.1 | 2 - 3 | 666 | 65 | ||
| XM_021249501.1 | XP_021105160.1 | 2 - 3 | 666 | 65 | ||
| XM_021249502.1 | XP_021105161.1 | 3 - 4 | 442 | 65 | ||
| XM_021249503.1 | XP_021105162.1 | 2 - 3 | 666 | 65 | ||
More Information
Set Membership: |
|

Sh2d2a
Genomic Map