Assay Details
| Gene Symbol: Sipa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004852386.3 | XP_004852443.1 | 10 - 11 | 2704 | 57 | |
| XM_013074954.2 | XP_012930408.1 | 10 - 11 | 2704 | 57 | ||
| XM_021264591.1 | XP_021120250.1 | 11 - 12 | 2785 | 57 | ||
| XM_021264592.1 | XP_021120251.1 | 10 - 11 | 3419 | 57 | ||
More Information
Set Membership: |
|

Sipa1
Genomic Map