Assay Details
| Gene Symbol: C1qa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004850533.2 | XP_004850590.1 | 2 - 3 | 497 | 89 | |
| XM_004850534.3 | XP_004850591.1 | 2 - 3 | 361 | 89 | ||
| XM_004850535.3 | XP_004850592.1 | 2 - 3 | 381 | 89 | ||
More Information
Set Membership: |
|

C1qa
Genomic Map