Assay Details
| Gene Symbol: Cd86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004873593.3 | XP_004873650.2 | 6 - 7 | 1041 | 86 | |
| XM_021251280.1 | XP_021106939.1 | 6 - 7 | 1145 | 86 | ||
More Information
Set Membership: |
|

Cd86
Genomic Map