Assay Details
| Gene Symbol: Cdk8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854819.3 | XP_004854876.1 | 4 - 5 | 502 | 70 | |
| XM_004854820.3 | XP_004854877.1 | 4 - 5 | 502 | 70 | ||
| XM_004854821.3 | XP_004854878.1 | 4 - 5 | 502 | 70 | ||
| XM_004854822.3 | XP_004854879.1 | 4 - 5 | 502 | 70 | ||
More Information
Set Membership: |
|

Cdk8
Genomic Map