Assay Details
| Gene Symbol: Nemp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004861263.3 | XP_004861320.1 | 2 - 3 | 272 | 72 | |
| XM_021241424.1 | XP_021097083.1 | 3 - 4 | 407 | 72 | ||
More Information
Set Membership: |
|

Nemp1
Genomic Map