Assay Details
| Gene Symbol: Tbxt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004856932.2 | XP_004856989.1 | 7 - 8 | 1188 | 60 | |
| XM_013076690.2 | XP_012932144.1 | 7 - 8 | 1191 | 60 | ||
More Information
Set Membership: |
|

Tbxt
Genomic Map