Assay Details
| Gene Symbol: Cant1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004860841.3 | XP_004860898.2 | 2 - 3 | 1430 | 52 | |
| XM_004860842.3 | XP_004860899.2 | 2 - 3 | 854 | 52 | ||
| XM_004860843.3 | XP_004860900.2 | 2 - 3 | 766 | 52 | ||
More Information
Set Membership: |
|

Cant1
Genomic Map