Assay Details
| Gene Symbol: Slc39a13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013075009.2 | XP_012930463.1 | 5 - 6 | 718 | 67 | |
| XM_021264197.1 | XP_021119856.1 | 5 - 6 | 718 | 67 | ||
| XM_021264198.1 | XP_021119857.1 | 5 - 6 | 767 | 67 | ||
| XM_021264199.1 | XP_021119858.1 | 5 - 6 | 1237 | 67 | ||
| XM_021264200.1 | XP_021119859.1 | 5 - 6 | 718 | 67 | ||
| XM_021264201.1 | XP_021119860.1 | 5 - 6 | 718 | 67 | ||
| XM_021264202.1 | XP_021119861.1 | 5 - 6 | 718 | 67 | ||
| XM_021264203.1 | XP_021119862.1 | 5 - 6 | 718 | 67 | ||
| XM_021264204.1 | XP_021119863.1 | 5 - 6 | 718 | 67 | ||
More Information
Set Membership: |
|

Slc39a13
Genomic Map