Assay Details
| Gene Symbol: Cdkn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021262037.1 | XP_021117696.1 | 8 - 9 | 870 | 86 | |
| XM_021262041.1 | XP_021117700.1 | 8 - 9 | 867 | 86 | ||
| XM_021262042.1 | XP_021117701.1 | 8 - 9 | 867 | 86 | ||
| XM_021262045.1 | XP_021117704.1 | 7 - 8 | 825 | 86 | ||
| XM_021262050.1 | XP_021117709.1 | 8 - 9 | 903 | 86 | ||
| XM_021262057.1 | XP_021117716.1 | 7 - 8 | 872 | 86 | ||
| XM_021262062.1 | XP_021117721.1 | 7 - 8 | 738 | 86 | ||
| XM_021262073.1 | XP_021117732.1 | 7 - 8 | 735 | 86 | ||
More Information
Set Membership: |
|

Cdkn3
Genomic Map