Assay Details
| Gene Symbol: Syt12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021264523.1 | XP_021120182.1 | 6 - 7 | 1422 | 66 | |
| XM_021264524.1 | XP_021120183.1 | 7 - 8 | 1551 | 66 | ||
| XM_021264525.1 | XP_021120184.1 | 8 - 9 | 1647 | 66 | ||
| XM_021264526.1 | XP_021120185.1 | 7 - 8 | 1518 | 66 | ||
More Information
Set Membership: |
|

Syt12
Genomic Map