Assay Details
| Gene Symbol: Repin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004860449.3 | XP_004860506.1 | 2 - 3 | 1439 | 75 | |
| XM_004860450.3 | XP_004860507.1 | 2 - 3 | 298 | 75 | ||
| XM_021240877.1 | XP_021096536.1 | 2 - 3 | 560 | 75 | ||
| XM_021240878.1 | XP_021096537.1 | 3 - 4 | 590 | 75 | ||
| XM_021240879.1 | XP_021096538.1 | 2 - 3 | 322 | 75 | ||
| XM_021240880.1 | XP_021096539.1 | 2 - 3 | 252 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004860450.3 transcripts.
Set Membership: |
|

Repin1
Genomic Map