Assay Details
| Gene Symbol: Aktip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847919.3 | XP_004847976.1 | 3 - 4 | 487 | 59 | |
| XM_013073333.2 | XP_012928787.1 | 3 - 4 | 419 | 59 | ||
| XM_021260903.1 | XP_021116562.1 | 3 - 4 | 460 | 59 | ||
| XM_021260904.1 | XP_021116563.1 | 3 - 4 | 354 | 59 | ||
More Information
Set Membership: |
|

Aktip
Genomic Map