Assay Details
| Gene Symbol: Slc1a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004848608.3 | XP_004848665.1 | 5 - 6 | 844 | 71 | |
| XM_021261494.1 | XP_021117153.1 | 3 - 4 | 311 | 71 | ||
More Information
Set Membership: |
|

Slc1a3
Genomic Map