Assay Details
| Gene Symbol: Gucy1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004868550.3 | XP_004868607.2 | 3 - 4 | 782 | 98 | |
| XM_004868551.3 | XP_004868608.1 | 3 - 4 | 741 | 98 | ||
| XM_013067011.2 | XP_012922465.1 | 3 - 4 | 465 | 98 | ||
| XM_021247445.1 | XP_021103104.1 | 3 - 4 | 748 | 98 | ||
| XM_021247446.1 | XP_021103105.1 | 3 - 4 | 750 | 98 | ||
More Information
Set Membership: |
|

Gucy1a1
Genomic Map