Assay Details
| Gene Symbol: Gucy1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004868550.3 | XP_004868607.2 | 8 - 9 | 2330 | 63 | |
| XM_004868551.3 | XP_004868608.1 | 8 - 9 | 2289 | 63 | ||
| XM_013067011.2 | XP_012922465.1 | 8 - 9 | 2013 | 63 | ||
| XM_021247447.1 | XP_021103106.1 | 7 - 8 | 1749 | 63 | ||
More Information
Set Membership: |
|

Gucy1a1
Genomic Map