Assay Details
| Gene Symbol: Chst1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004851971.3 | XP_004852028.1 | 3 | 6418 | 80 | |
| XM_013074974.2 | XP_012930428.1 | 4 | 1279 | 80 | ||
| XM_013074975.2 | XP_012930429.1 | 4 | 3100 | 80 | ||
| XM_021264297.1 | XP_021119956.1 | 2 | 6824 | 80 | ||
| XM_021264298.1 | XP_021119957.1 | 3 | 1685 | 80 | ||
| XM_021264299.1 | XP_021119958.1 | 3 | 6605 | 80 | ||
| XM_021264300.1 | XP_021119959.1 | 3 | 3506 | 80 | ||
More Information
Set Membership: |
|

Chst1
Genomic Map