Assay Details
| Gene Symbol: Cpt1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845287.3 | XP_004845344.1 | 4 - 5 | 645 | 56 | |
| XM_004845288.3 | XP_004845345.1 | 4 - 5 | 657 | 56 | ||
| XM_004845289.3 | XP_004845346.1 | 4 - 5 | 625 | 56 | ||
| XM_021258706.1 | XP_021114365.1 | 3 - 4 | 475 | 56 | ||
More Information
Set Membership: |
|

Cpt1b
Genomic Map